Learn More
Erlab™ Captair by Erlab™ PVC Glove Box
Product Code. 15170813
Click to view available options
Material:
PVC
Unit Size:
Each
Product Code. 15170813
Brand: Erlab™ 2200ANMXL
Specifications
Product Type Glove Box
Material PVC
Depth (Metric) Interior 560 mm
Width (Metric) Interior 860 mm