Learn More
Invitrogen™ CAPG Recombinant Rabbit Monoclonal Antibody (24GB3595)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556998
This item is not returnable.
View return policy
Description
CAPG Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a member of the gelsolin/villin family of actin-regulatory proteins. The encoded protein reversibly blocks the barbed ends of F-actin filaments in a Ca2+ and phosphoinositide-regulated manner, but does not sever preformed actin filaments. By capping the barbed ends of actin filaments, the encoded protein contributes to the control of actin-based motility in non-muscle cells. Alternatively spliced transcript variants have been observed, but have not been fully described.
Specifications
| CAPG | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CAPG | |
| Actin regulatory protein CAP-G; actin-capping protein GCAP39; actin-regulatory protein CAP-G; AFCP; Capg; capping actin protein, gelsolin like; capping protein (actin filament), gelsolin-like; epididymis secretory protein Li 66; gCap39; gelsolin-like capping protein; HEL-S-66; macrophage capping protein; macrophage-capping protein; mbh1; MCP; myc basic motif homolog 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12332, 297339, 822 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3595 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P24452, P40121, Q6AYC4 | |
| CAPG | |
| A synthesized peptide derived from human CAPG (aa 1-188). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |