Learn More
Candesartan-d4, MedChemExpress
Description
Candesartan D4 (CV-11974 D4) is the deuterium labeled Candesartan, which is an angiotensin II receptor antagonist.
Specifications
Specifications
| Chemical Name or Material | Candesartan-d4 |
| CAS | 1346604-70-3 |
| Purity Grade Notes | Research |
| Physical Form | Solid |
| Molecular Formula | C24H16D4N6O3 |
| Color | White |
| Synonym | CV-11974-d4 |
| SMILES | [2H]C1=C(CN2C(C(C(O)=O)=CC=C3)=C3N=C2OCC)C([2H])=C([2H])C(C4=CC=CC=C4C5=NNN=N5)=C1[2H] |
| Molecular Weight (g/mol) | 444.48 |
| Quantity | 10 mg |
| Show More |