Learn More
CAMK2B Monoclonal Antibody (OTI4B12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808428S
This item is not returnable.
View return policy
Description
CAMK2B Monoclonal antibody specifically detects CAMK2B in Human samples. It is validated for Western BlotSpecifications
| CAMK2B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P41279 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1326 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4B12 | |
| Unconjugated | |
| MAP3K8 | |
| Ca++/calmodulin-dependent protein kinase II beta subunit; calcium/calmodulin dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II, beta; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II subunit beta; CaM kinase II beta subunit; caM kinase II subunit beta; CAM2; CaMK II; CAMK2; CAMK2B; Camk2d; CAMKB; caMK-II subunit beta; caM-kinase II beta chain; Ck2b; hypothetical protein LOC525416; MGC29528; proline rich calmodulin-dependent protein kinase; β CaMKII; βCaMKII | |
| Human recombinant protein fragment corresponding to amino acids 277-461 of human MAP3K8 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |