missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAMK2B Monoclonal Antibody (OTI4B12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808428
This item is not returnable.
View return policy
Description
The Ca2+/calmodulin-dependent protein kinases (CaM kinases) comprise a structurally related subfamily of serine/threonine kinases which include CaMKI, CaMKII and CaMKIV. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+ and calmodulin (CaM) and has been implicated in regulation of the cell cycle and transcription. There are four CaMKII isozymes, designated alpha, beta, gamma and delta, which may or may not be co-expressed in the same tissue type. CaMKII alpha is autophosphorylated on Thr 286 upon the binding of the Ca2+/CaM complex to the autoinhibitory domain of CaMKII. This process is called Ca2+/CaM trapping, which is thought to be involved in the synaptic encoding of information.Specifications
| CAMK2B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13554 | |
| CAMK2B | |
| Human recombit protein fragment corresponding to amino acids 311-385 of human CAMK2B produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4B12 | |
| Unconjugated | |
| CAMK2B | |
| CAM2, CAMK2, CAMKB, MRD54 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 816 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction