missing translation for 'onlineSavingsMsg'
Learn More

Camicinal, MedChemExpress

Product Code. p-200097700
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477007 5 mg 5mg
16470617 50 mg 50mg
16464288 10 mg 10mg
16435216 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16477007 Supplier Medchem Express Supplier No. HY10922

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Camicinal (GSK962040) is a small molecule, selective motilin receptor agonist with pEC50 of 7.9.

Camicinal (GSK962040) is a small molecule, selective motilin receptor agonist with pEC50 of 7.9.

Specifications

Chemical Name or Material Camicinal
CAS 923565-21-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H33FN4O
Quantity 5 mg
Synonym GSK962040
Solubility Information DMSO : 100 mg/mL (235.54 mM; Need ultrasonic)
SMILES O=C(N1CCC(NC2=CC=CC(F)=C2)CC1)CC3=CC=C(CN4C[C@H](C)NCC4)C=C3
Molecular Weight (g/mol) 424.55
Formula Weight 424.55
Percent Purity 95.94%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.