missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride Anhydrous MP Biomedicals

Product Code. 11448801 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11448801

Brand: MP Biomedicals 0215350280

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 10043-52-4
Quantity 100 g
Molecular Formula CaCl2
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Ca++]
IUPAC Name calcium dichloride
Molecular Weight (g/mol) 110.98
PubChem CID 5284359
ChEBI CHEBI:3312
Formula Weight 147.02 (110.99 Anhydrous)
Density 1.34 g/cm3 at 20 °C
Chemical Name or Material Calcium Chloride, Anhydrous
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.