missing translation for 'onlineSavingsMsg'
Learn More

Melford Calcium Chloride 2M Solution

Product Code. 30018137
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
This item is not returnable. View return policy

Product Code. 30018137

Brand: Melford C2510050.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Supplied as 5 x 10 Milliliters.

Supplied as 5 x 10 mm. Sterile filtered to 0.22 μm

Specifications

Chemical Name or Material Calcium Chloride 2 M Solution
CAS 10035-04-8, 7732-18-5
Molecular Formula CaCl2
Quantity 50 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.