missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Calcein tetraethyl ester

Product Code. 16414687
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16414687 50 mg 50mg
16488906 100 mg 100mg
16438768 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16414687 Supplier MedChemExpress Supplier No. HYD0830

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Calcein tetraethyl ester is a fluorescent dye, is used in biology as it can be transported through the cellular membrane into live cells, which makes it useful for testing of cell viability and for short-term labeling of cells.

Specifications

Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Product Type Dye Reagents
Quantity 50 mg
Shipping Condition Room temperature
Solubility DMSO : 25 mg/mL (34.03 mM; Need ultrasonic) ∣ H2O : < 0.1 mg/mL (insoluble)
Target HBV
Molecular Weight (g/mol) 734.75 g/mol
CAS 1170856-93-5
Synonym Fluorexon tetraethyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.