missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAGE1 Polyclonal Antibody
Description
CAGE1 Polyclonal Antibody detects endogenous levels of CAGE1 protein.
CAGE1 (Cancer Antigen 1) is a Protein Coding gene. Diseases associated with CAGE1 include Fat Necrosis Of Breast and Amelogenesis Imperfecta, Type If. [GeneCards].
Specifications
Specifications
| Antigen | CAGE1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CAGE1 |
| Gene Accession No. | Q8TC20 |
| Gene Alias | 4933427I01Rik; bA69L16.7; CAGE1; CAGE-1; cancer antigen 1; cancer/testis antigen 3; Cancer/testis antigen 3 homolog; cancer/testis antigen 95; cancer/testis antigen gene 1; cancer-associated gene 1 protein; Cancer-associated gene 1 protein homolog; CT3; CT3 homolog; CT95; Ctag3 |
| Gene Symbols | CAGE1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?