Learn More
Cadence Science™ Non-Coring Septum Penetration Needles
Click to view available options
Needle Gauge:
18 G
20 G
Length (English) Needle:
1 in.
6 in.
8 in.
12 in.
Unit Size:
Pack of 12
Description
Needles allow for easier penetration and minimize coring and tracking
- Micro-mate 316 stainless-steel hub
- 316 stainless-steel tubing
Specifications
Specifications
| Material | Stainless Steel (Hub) |
| Needle Gauge | 20 G |
| For Use With (Application) | For easier penetration and minimize coring and tracking |
| Length (English) Needle | 1 in. |
| Product Type | Penetration Needle |
| Quantity | 12 Needles |
| Length (Metric) Needle | 25 mm |
| Needle Point Style | Deflected End |