missing translation for 'onlineSavingsMsg'
Learn More

Cacng2/Cacng4/Cacng8 Mouse anti-Rat, FITC, Clone:S245-36, Abnova™

Product Code. 16119869
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16119869 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16119869 Supplier Abnova Supplier No. MAB17336.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against partial recombinant rat Cacng2/Cacng4/Cacng8.

Specifications

Antigen Cacng2/Cacng4/Cacng8
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone S245-36
Conjugate FITC
Description calcium channel, voltage-dependent, gamma subunit 2
Dilution Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Cacng2
Gene Alias Ipr328
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 203-323 at C-terminus of rat Cacng2.
Quantity 100 μg
Gene ID (Entrez) 84347, 140725, 140729
Target Species Rat
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.