missing translation for 'onlineSavingsMsg'
Learn More

CaCCinh-A01, MedChemExpress

Product Code. 16464396
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464396 100 mg 100mg
16493098 10 mg 10mg
16469656 50 mg 50mg
16425837 5 mg 5mg
16422626 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16464396 Supplier Medchem Express Supplier No. HY100611

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CaCCinh-A01 is an inhibitor of both TMEM16A and calcium-activated chloride channel (CaCC) with IC50s of 2.1 and 10 μM, respectively.

CaCCinh-A01 is an inhibitor of both TMEM16A and calcium-activated chloride channel (CaCC) with IC50s of 2.1 and 10 μM, respectively.

Specifications

Chemical Name or Material CaCCinh-A01
Physical Form Solid
CAS 407587-33-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H21NO4S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 50 mg/mL (143.91 mM)
SMILES O=C(C1=C(NC(C2=CC=CO2)=O)SC3=C1CCC(C(C)(C)C)C3)O
Molecular Weight (g/mol) 347.43
Formula Weight 347.43
Percent Purity 99.79%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.