missing translation for 'onlineSavingsMsg'
Learn More

Cabozantinib S-malate, MedChemExpress

Product Code. 16467687
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467687 5 mg 5mg
16481026 200 mg 200mg
16464948 10 mg 10mg
16445726 100 mg 100mg
16431197 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16467687 Supplier Medchem Express Supplier No. HY12044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cabozantinib S-malate (XL184 S-malate) is a potent multiple receptor tyrosine kinases inhibitor that inhibits VEGFR2, c-Met, Kit, Axl and Flt3 with IC50s of 0.035, 1.3, 4.6, 7 and 11.3 nM, respectively.

Cabozantinib S-malate (XL184 S-malate) is a potent multiple receptor tyrosine kinases inhibitor that inhibits VEGFR2, c-Met, Kit, Axl and Flt3 with IC50s of 0.035, 1.3, 4.6, 7 and 11.3 nM, respectively.

Specifications

Chemical Name or Material Cabozantinib S-malate
CAS 1140909-48-3
Color White
Purity Grade Notes Research
Molecular Formula C32H30FN3O10
Quantity 5 mg
Synonym XL184 S-malateBMS-907351 S-malate
Solubility Information DMSO : ≥ 23 mg/mL (36.19 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C([C@H](CC(O)=O)O)O.O=C(NC1=CC=C(C=C1)OC2=CC=NC3=CC(OC)=C(C=C23)OC)C4(CC4)C(NC5=CC=C(C=C5)F)=O
Molecular Weight (g/mol) 635.59
Formula Weight 635.59
Percent Purity 99.29%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.