missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cabozantinib, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Description
Cabozantinib is a potent and orally active inhibitor of VEGFR2 and MET, with IC50 values of 0.035, and 1.3 nM, respectively. Cabozantinib displays strong inhibition of KIT, RET, AXL, TIE2, and FLT3 (IC50=4.6, 5.2, 7, 14.3, and 11.3 nM, respectively). Cabozantinib shows antiangiogenic activity. Cabozantinib disrupts tumor vasculature and promotes tumor and endothelial cell apoptosis.
Specifications
Specifications
| Chemical Name or Material | Cabozantinib |
| CAS | 849217-68-1 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C28H24FN3O5 |
| Quantity | 200 mg |
| Synonym | XL184 BMS-907351 |
| Solubility Information | DMSO : ≥ 30 mg/mL (59.82 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | O=C(C1(CC1)C(NC2=CC=C(F)C=C2)=O)NC3=CC=C(C=C3)OC4=C5C=C(OC)C(OC)=CC5=NC=C4 |
| Molecular Weight (g/mol) | 501.51 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction