Learn More
asecos™ Cabinet for Chemicals CS-CLASSIC
Product Code. p-7163620 Shop All asecos Products
Click to view available options
Configuration:
2 Shelves, 1 Perforated Insert, 1 Bottom Collecting Sump
3 Pull-out Shelves
3 Shelves, 1 Perforated Insert, 1 Bottom Collecting Sump
4 Pull-out Shelves
6 Pull-out Shelves
Color:
Anthracite Gray, Light Gray
Anthracite Gray, Warning Yellow
Width (Metric) Exterior:
810 mm
1055 mm
Unit Size:
Each
For indoor use only
Product Code. 15915006
Brand: asecos™ 3053004130534
This item requires a price update prior to being ordered.
Request a Price for this item here.
For indoor use only
Safe and proper storage of non-flammable chemicals and toxic substances in working areas
  • Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated textured surface
  • No unauthorised use: doors lockable with cylinder locking (integrated in the turning handle)
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: natural ventilation due to ventilation openings at the cabinet bottom, ready for connection to a technical exhaust system
Specifications
Certifications/Compliance DIN EN 16121/16122 conformity
Configuration 2 Shelves, 1 Perforated Insert, 1 Bottom Collecting Sump
Depth (Metric) Exterior 520 mm
Depth (Metric) Interior 493 mm
Dimensions (D x W x H) Interior 493 x 751 x 990 mm
Dimensions (D x W x H) 520 x 810 x 1105 mm
Door Style Wing
For Use With (Application) Safe and proper storage of non-flammable chemicals and toxic substances in working areas
Height (Metric) Exterior 1105 mm
Height (Metric) Interior 990 mm
Load Bearing Capacity 60 kg Shelves
Model CS.110.081
No. of Doors 2
No. of Shelves 2
Shelf Type Standard
Surface Coating Powder-coated textured
Weight (Metric) 67 kg
Width (Metric) Exterior 810 mm
Width (Metric) Interior 751 mm
Color Anthracite Gray, Light Gray
Depth (English) Exterior 20.47 in.
Height (English) Exterior 43.50 in.
Height (Metric) Opening 990 mm
Material Steel
Product Type Chemical Cabinet
Width (English) Exterior 31.89 in.