missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA-170, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
Description
CA-170 is an orally delivered dual inhibitor of VISTA and PD-L1. CA-170 exhibits potent rescue of proliferation and effector functions of T cells inhibited by PD-L1/L2 and VISTA with selectivity over other immune checkpoint proteins as well as a broad panel of receptors and enzymes.
Specifications
Specifications
| Chemical Name or Material | CA-170 |
| Physical Form | Solid |
| CAS | 1673534-76-3 |
| Color | Light Yellow |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C12H20N6O7 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : 28.5 mg/mL (79.10 mM; Need ultrasonic and warming) |
| SMILES | C[C@@H](O)[C@@H](C(O)=O)NC(N[C@H](C1=NC([C@@H](N)CO)=NO1)CC(N)=O)=O |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction