missing translation for 'onlineSavingsMsg'
Learn More

CA-074 methyl ester, MedChemExpress

Product Code. 16462958
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16462958 10 mg 10mg
16485697 5 mg 5mg
16452556 25 mg 25mg
16420639 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16462958 Supplier Medchem Express Supplier No. HY100350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CA-074 methyl ester is a specific inhibitor of Cathepsin B, which has potent bioactivities such as neuroprotective, anti-cancer, and anti-inflamatory effects.

CA-074 methyl ester is a specific inhibitor of Cathepsin B, which has potent bioactivities such as neuroprotective, anti-cancer, and anti-inflamatory effects.

Specifications

Chemical Name or Material CA-074 methyl ester
CAS 147859-80-1
Color White
Purity Grade Notes Research
Molecular Formula C19H31N3O6
Quantity 10 mg
Synonym CA-074Me
Solubility Information DMSO : 100 mg/mL (251.59 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C([C@@H]1[C@@H](C(NCCC)=O)O1)N[C@]([C@@H](C)CC)([H])C(N(CCC2)[C@@H]2C(OC)=O)=O
Molecular Weight (g/mol) 397.47
Formula Weight 397.47
Percent Purity 98.2%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.