missing translation for 'onlineSavingsMsg'
Learn More

C7orf10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. p-7209934 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16831195 20 μL 20µL
16821195 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16831195 Supplier Proteintech Supplier No. 215891AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Russel-Silver syndrome candidate,dermal papilla derived protein 13,hypothetical protein LOC79783

Specifications

Antigen C7orf10
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SUGCT
Gene Accession No. Q68FU4, Q7TNE1, Q9HAC7
Gene Alias C7orf10, DERP13, ORF19
Gene Symbols Sugct
Host Species Rabbit
Immunogen C7orf10 Fusion Protein Ag16159
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192136, 361253, 79783
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.