Description
- Bamboo-TwinFlex™ Technology ensures cut protection and abrasion resistance
- Bamboo fiber is soft and silky for increased comfort
- Knit cuff offers a secure fit
- Silicone-free according to the imprint test
- Ideal for applications in the automotive, construction, beverage, glass, and metal industries
Specifications
Specifications
| Size | Large |
| Product Type | Cut-resistant Sleeve |
| Material | Bamboo Viscose, HPPE, Polyamide |
| Color | Green |
| Sterility | Non-sterile |
| Style | Standard Sleeve |
| Disposable | Reusable |
| Closure Type | Elastic Ends |
| Seam Type | Taped |
| Cuff Style | Knit |