missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C18orf54 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531692
This item is not returnable.
View return policy
Description
Recommended positive controls: NT2D1, SK-N-SH. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Might play a role in cell proliferation.
Specifications
| C18orf54 | |
| Polyclonal | |
| Unconjugated | |
| C18orf54 | |
| C18orf54; chromosome 18 open reading frame 54; LAS2; Lung adenoma susceptibility protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 162681 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.69 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8IYD9 | |
| C18orf54 | |
| Recombinant protein encompassing a sequence within the center region of human C18orf54. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction