Learn More
Eurofins DiscoverX c-Kit (D816V) Protein, Active
Description
Binding of stem cell factor (SCF) to C-Kit receptor tyrosine kinase induces homodimerization and intermolecular tyrosine phosphorylation of the receptor, creating docking sites for a number of SH2-containing signal transduction molecules. These include phosphatidylinositol 3'-kinase (PI 3K) and phospholipase C-1, Stat1, the Src family kinases Src, Lyn, and Fyn, and the GTPase activating protein GAP. Mutation in Kit which result in ligand-independent activation of the kinase were found to be associated with human gastrointestinal stromal tumors and mastocytosis.
GenBank X06182
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 76.7 kDa |
| Quantity | 250 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal GST |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥79% |
| Protein | c-Kit (D816V), active |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.