missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX c-Kit (D816V) Protein, Active

Product Code. 17360424
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17360424 10 μg 10µg
17370424 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17360424 Supplier Eurofins DiscoverX Supplier No. 14611

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-terminal GST tagged, recombinant human c-Kit, amino acids 544-end containing the D816V mutation, for use in Kinase Assays.

Binding of stem cell factor (SCF) to C-Kit receptor tyrosine kinase induces homodimerization and intermolecular tyrosine phosphorylation of the receptor, creating docking sites for a number of SH2-containing signal transduction molecules. These include phosphatidylinositol 3'-kinase (PI 3K) and phospholipase C-1, Stat1, the Src family kinases Src, Lyn, and Fyn, and the GTPase activating protein GAP. Mutation in Kit which result in ligand-independent activation of the kinase were found to be associated with human gastrointestinal stromal tumors and mastocytosis.

GenBank X06182

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 76.7 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal GST
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Truncated
Purity or Quality Grade ≥79%
Protein c-Kit (D816V), active
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.