missing translation for 'onlineSavingsMsg'
Learn More

Butein, MedChemExpress

Product Code. 16470276
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470276 2 mg 2mg
16489617 5 mg 5mg
16463117 50 mg 50mg
16457116 100 mg 100mg
16437078 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16470276 Supplier Medchem Express Supplier No. HY16558

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Butein is a cAMP-specific PDE inhibitor with an IC50 of 10.4 μM for PDE4. Butein is a specific protein tyrosine kinase inhibitor with IC50s of 16 and 65 μM for EGFR and p60c-src in HepG2 cells. Butein sensitizes HeLa cells to Cisplatin through AKT and ERK/p38 MAPK pathways by targeting FoxO3a. Butein is a SIRT1 activator (STAC).

Butein is a cAMP-specific PDE inhibitor with an IC50 of 10.4 μM for PDE4. Butein is a specific protein tyrosine kinase inhibitor with IC50s of 16 and 65 μM for EGFR and p60c-src in HepG2 cells. Butein sensitizes HeLa cells to Cisplatin through AKT and ERK/p38 MAPK pathways by targeting FoxO3a. Butein is a SIRT1 activator (STAC).

Specifications

Chemical Name or Material Butein
CAS 487-52-5
Color Brownish Yellow
Purity Grade Notes Research
Molecular Formula C15H12O5
Quantity 2 mg
Synonym 2’,3,4,4’-tetrahydroxy Chalcone
Solubility Information DMSO : 50 mg/mL (183.65 mM; Need ultrasonic)
SMILES OC1=CC=C(C(/C=C/C2=CC(O)=C(O)C=C2)=O)C(O)=C1
Molecular Weight (g/mol) 272.25
Formula Weight 272.25
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.