Learn More
Busulfan, MedChemExpress
Product Code. 16430848
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16430848 5 g 5g
16462109 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16430848 Supplier Medchem Express Supplier No. HYB0245
MedChemExpress Busulfan is a potent alkylator with selective immunosuppressive effect on bone marrow.
Busulfan is a potent alkylator with selective immunosuppressive effect on bone marrow.
Specifications
Chemical Name or Material Busulfan
CAS 55-98-1
Color White
Purity Grade Notes Research
Molecular Formula C6H14O6S2
Quantity 5 g
Solubility Information DMSO : 50 mg/mL (203.00 mM; Need ultrasonic) ∣H2O : 1 mg/mL (4.06 mM; ultrasonic and warming and heat to 80°C)
SMILES CS(=O)(OCCCCOS(C)(=O)=O)=O
Molecular Weight (g/mol) 246.3
Formula Weight 246.3
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease