missing translation for 'onlineSavingsMsg'
Learn More

Buquinolate, TRC

Product Code. 30134557
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134557 50 mg 50mg
17908384 5 mg 5mg
17950015 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134557 Supplier Toronto Research Chemicals Supplier No. TRCB83583550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5486-03-3
Molecular Formula C20 H27 N O5
Molecular Weight (g/mol) 361.43
Synonym 3-Quinolinecarboxylic acid, 4-hydroxy-6,7-diisobutoxy-, ethyl ester (7CI,8CI), 4-Hydroxy-6,7-diisobutoxyquinoline-3-carboxylic acid ethyl ester, Antagonal, Bonaid, Bonaid TM, Buquinolate, Butoril, Ethyl 4-hydroxy-6,7-bis(2-methylpropoxy)-3-quinolinecarboxylate, Ethyl 4-hydroxy-6,7-diisobutoxy-3-quinolinecarboxylate, Ethyl 6,7-diisobutoxy-4-hydroxyquinoline-3-carboxylate, NSC 201116, U 1093
IUPAC Name ethyl 4-hydroxy-6,7-bis(2-methylpropoxy)quinoline-3-carboxylate
SMILES CCOC(=O)c1cnc2cc(OCC(C)C)c(OCC(C)C)cc2c1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C20H27NO5/c1-6-24-20(23)15-9-21-16-8-18(26-11-13(4)5)17(25-10-12(2)3)7-14(16)19(15)22/h7-9,12-13H,6,10-11H2,1-5H3,(H,21,22)
Formula Weight 361.1889
Chemical Name or Material Buquinolate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.