missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bupivacaine Hydrochloride, 100.3%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Color | White |
| Assay Percent Range | 100.3% |
| Quantity | 1 g |
| Formula Weight | 324.9 (Anhydrous) |
| Physical Form | Powder |
| Chemical Name or Material | Bupivacaine Hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction