Learn More
Buffer Solution pH5.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Product Code. 10294430 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
500mL
Product Code. 10294430
Brand: Fisher Chemical J/2830/17
Request Bulk or Custom Format
Buffer Solution pH5.00 (Phthalate), NIST Standard Solution ready to use for pH measurement
TRUSTED_SUSTAINABILITY
Specifications
CAS 877-24-7
Color Colorless
pH 5
Physical Form Liquid
Quantity 2.5 L
MDL Number 13070
Packaging HDPE plastic bottle