Learn More
Buffer concentrate, pH 1.00, hydrochloric acid / potassium chloride, Honeywell Fluka™
Product Code. 15625320 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 Pc.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15625320 1 Pc. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15625320 Supplier Honeywell-Fluka Supplier No. 387401EA
Request Bulk or Custom Format
for 500 ml buffer solution, hydrochloric acid / potassium chloride
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Buffer concentrate
pH 1
Quantity 1 Pc.