Learn More
Buffer Colour Coded Concentrated Solution pH4.00 (Phthalate) Red, NIST Standard for pH measurement, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 Ampule
6 Ea.
Unit Size:
1 ampoule
Pack of 6
Specifications
Specifications
| Color | Red |
| Physical Form | Liquid |
| Quantity | 6 Ea. |
| MDL Number | 217672 |
| Packaging | Ampoule |