Learn More
Bucumolol, TRC
Product Code. 17997464
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17997464 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17997464 Supplier Toronto Research Chemicals Supplier No. TRCB6893952.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 58409-59-9
Molecular Formula C17H23NO4
Molecular Weight (g/mol) 305.37
Synonym 8-[3-[(1,1-Dimethylethyl)amino]-2-hydroxypropoxy]-5-methyl-2H-1-benzopyran-2-one; dl-Bucumolol
IUPAC Name 8-[3-(tert-butylamino)-2-hydroxypropoxy]-5-methylchromen-2-one
SMILES Cc1ccc(OCC(O)CNC(C)(C)C)c2OC(=O)C=Cc12
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C17H23NO4/c1-11-5-7-14(16-13(11)6-8-15(20)22-16)21-10-12(19)9-18-17(2,3)4/h5-8,12,18-19H,9-10H2,1-4H3
Formula Weight 305.1627
Chemical Name or Material Bucumolol