missing translation for 'onlineSavingsMsg'
Learn More

BTN3A1 Mouse anti-Human, Clone:4E1B9, Abnova™

Product Code. 16035296
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16035296 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16035296 Supplier Abnova Supplier No. MAB18003.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against partial recombinant human BTN3A1.

Specifications

Antigen BTN3A1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4E1B9
Conjugate Unconjugated
Description butyrophilin, subfamily 3, member A1
Dilution ELISA (1:10000)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene BTN3A1
Gene Alias BT3.1, BTF5, CD277, MGC141880
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 30-254 of human BTN3A1.
Quantity 100 μg
Gene ID (Entrez) 11119
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.