Learn More
BTG2 Monoclonal Antibody (OTI2G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808151
Encompass_Preferred
 View more versions of this product
Product Code. 17539093
  • 540.87 EUR / 100µL
Description
The protein encoded by this gene is a member of the BTG/Tob family. This family has structurally related proteins that appear to have antiproliferative properties. This encoded protein is involved in the regulation of the G1/S transition of the cell cycle.
Specifications
Specifications
BTG2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P78543
BTG2
Full length human recombit protein of human BTG2 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI2G5
Unconjugated
BTG2
APRO1, PC3, TIS21
Mouse
Affinity Chromatography
RUO
7832
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions