Learn More
Paragon Scientific™ BS/U-tube Viscometer for Transparent Liquids
Click to view available options
Viscosity Range:
0.9 to 3 cSt
2.0 to 10 cSt
20 to 100 cSt
200 to 1000 cSt
2000 to 10,000 cSt
6 to 30 cSt
60 to 300 cSt
600 to 3000 cSt
Unit Size:
Each
Description
- High Quality glass capillary viscometer
- Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
Specifications
| Viscosity Range | 2000 to 10,000 cSt |
| Calibrated | No |
| Approximate Constant | 10 |
| ASTM Numbers | D 445, D 446 |
| Certifications/Compliance | ASTM D 445, D 446 and ISO 3104, 3105 |
| Size | H |
4 week lead-time