Learn More
Cole Parmer Brushes for Peristaltic Pump Drives
Product Code. 11827314
Change view
Click to view available options
Sterility:
Non-sterile
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility unitSize
11827314 Non-sterile Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11827314 Supplier Cole Parmer Supplier No. 0752004
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Brushes for Peristaltic Pump Drives are easy to install and help extend the life of a drive.
  • Quickly and easily replace components of your Masterflex system
  • Choose replacement brushes and caps based on Masterflex L/S drive
  • Brushes should be inspected every six months or 2000 hours, whichever comes first
Specifications
For Use With Masterflex L/S series 07520, 07521, 07522, 07523, 07525, 07526, 07527, 07528, 07553 (except -50, -60), 07554 (except -50, -60), 07558, 07567, 07568, 07592, 77300 drives
Sterility Non-sterile
Type Replacement brushes