Learn More
Cole Parmer Brushes for Peristaltic Pump Drives
Description
Brushes for Peristaltic Pump Drives are easy to install and help extend the life of a drive.
- Quickly and easily replace components of your Masterflex system
- Choose replacement brushes and caps based on Masterflex L/S drive
- Brushes should be inspected every six months or 2000 hours, whichever comes first
Specifications
Specifications
| For Use With | Masterflex L/S series 07520, 07521, 07522, 07523, 07525, 07526, 07527, 07528, 07553 (except -50, -60), 07554 (except -50, -60), 07558, 07567, 07568, 07592, 77300 drives |
| Sterility | Non-sterile |
| Type | Replacement brushes |