Learn More
BD Brucella Blood Agar with Hemin and Vitamin K1
Click to view available options
:
Brucella Blood Agar; Quantity: 20 Plates
Description
- BD Brucella Blood Agar with Hemin and Vitamin K1 is a modification of Brucella Agar that has been supplemented with hemin and vitamin K1 to support the growth of fastidious anaerobes, especially Bacteroides, Prevotella, and Porphyromonas when incubated anaerobically
- It is used for the isolation of strict anaerobes from clinical specimens.
- It is also used for susceptibility testing of anaerobes with the E test™ method.
Specifications
Specifications
| Media Type | Brucella Blood Agar |
| Formulation | Pancreatic Digest of Casein 10.0 g/L, Peptic Digest of Animal Tissue 10.0 g/L, Yeast Extract 2.0 g/L, Glucose 1.0 g/L, Sodium Chloride 5.0 g/L, Sodium Bisulfite 0.1 g/L, Hemin 0.005 g/L, Vitamin K1 0.01 g/L, Agar 15.0 g/L |
| No. per Pack | 20 |