Learn More
BD Brucella Blood Agar with Hemin and Vitamin K1
Product Code. 11922825
Click to view available options
:
Brucella Blood Agar; Quantity: 20 Plates
Product Code. 11922825
Brand: BD 255509
BD Brucella Blood Agar with Hemin and Vitamin K1 is a highly nutritious medium for the isolation and cultivation of strict anaerobes from clinical specimens.
  • BD Brucella Blood Agar with Hemin and Vitamin K1 is a modification of Brucella Agar that has been supplemented with hemin and vitamin K1 to support the growth of fastidious anaerobes, especially Bacteroides, Prevotella, and Porphyromonas when incubated anaerobically
  • It is used for the isolation of strict anaerobes from clinical specimens.
  • It is also used for susceptibility testing of anaerobes with the E test™ method.
TRUSTED_SUSTAINABILITY
Specifications
Media Type Brucella Blood Agar
Formulation Pancreatic Digest of Casein 10.0 g/L, Peptic Digest of Animal Tissue 10.0 g/L, Yeast Extract 2.0 g/L, Glucose 1.0 g/L, Sodium Chloride 5.0 g/L, Sodium Bisulfite 0.1 g/L, Hemin 0.005 g/L, Vitamin K1 0.01 g/L, Agar 15.0 g/L
No. per Pack 20