missing translation for 'onlineSavingsMsg'
Learn More

Bruceantin, MedChemExpress

Product Code. 16471528
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16471528

Brand: Medchem Express HYN0840

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bruceantin(NSC165563) is first isolated from Brucea antidysenterica, a tree used in Ethiopia for the treatment of cancer, and activity was observed against B16 melanoma, colon 38, and L1210 and P388 leukemia in mice.

Bruceantin(NSC165563) is first isolated from Brucea antidysenterica, a tree used in Ethiopia for the treatment of cancer, and activity was observed against B16 melanoma, colon 38, and L1210 and P388 leukemia in mice.

Specifications

Chemical Name or Material Bruceantin
CAS 41451-75-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C28H36O11
Quantity 5 mg
Synonym (-)-Bruceantin NCI165563 NSC165563
Solubility Information DMSO : 100 mg/mL (182.29 mM; Need ultrasonic)
SMILES O=C([C@@]1([C@H]([C@H](O)[C@]2([H])[C@]3(C4)C)O)[C@@]([C@H](C5=O)OC(/C=C(C)/C(C)C)=O)([H])[C@@]2([C@@](O5)([H])C[C@@]3([H])C(C)=C(O)C4=O)CO1)OC
Molecular Weight (g/mol) 548.58
Formula Weight 548.58
Percent Purity 96.57%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.