Learn More
VITLAB™ Brown Soda Lime Glass Reservoir Bottles
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Coating | Ethylene Acrylate |
| Color | Brown |
| For Use With (Application) | For volume measurement |
| Includes | Screw cap |
| Material | Soda Lime Glass |
| Max. Temperature (Metric) | 80°C |
| No. per Pack | 1Each |
| Operating Temperature | 80°C |
| Quantity | 1Each |
| Show More |