Learn More
Kautex™ Brown Low Density Polyethylene Wide Neck Bottles
Product Code. 10022283
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 10
Pack of 130
Pack of 20
Pack of 260
Pack of 4
Pack of 480
Pack of 50
Pack of 56
Pack of 650
Product Code. 10022283
Brand: Kautex™ 2000784585
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use brown-transparent Kautex™ Brown Low Density Polyethylene Wide Neck Bottles to limit the exposure to light and lessen possible tainting of bottle contents through light transmission.
  • Brown color protects light-sensitive materials against UV-light
  • Wide neck opening means bottles are easy to fill and clean
  • Round
  • With closure
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Color Brown
Diameter (Metric) Inner Neck 30.8 mm
Form Round
Material LDPE
Neck ID (Metric) 30.8 mm
No. per Case 260
Replacement Closure Screw Closure LDPE
Quantity 10 Pk.
Closure Included Yes
Graduated No
Height (Metric) 120 mm
Product Type Wide Neck Bottle
Closure Material LDPE
Closure Size 40 mm
Diameter (Metric) 64 mm
Closure Type Screw Cap