missing translation for 'onlineSavingsMsg'
Learn More

Brivanib (alaninate), MedChemExpress

Product Code. 16456557
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16456557

Brand: Medchem Express HY10336

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Brivanib alaninate (BMS-582664) is an ATP-competitive inhibitor against VEGFR2 with an IC50 of 25 nM; has moderate potency against VEGFR-1 and FGFR-1, but more than 240-fold against PDGFRβ.

Brivanib alaninate (BMS-582664) is an ATP-competitive inhibitor against VEGFR2 with an IC50 of 25 nM

Specifications

Chemical Name or Material Brivanib (alaninate)
CAS 649735-63-7
Color White
Purity Grade Notes Research
Molecular Formula C22H24FN5O4
Quantity 5 mg
Synonym BMS-582664
Solubility Information DMSO : 100 mg/mL (226.52 mM; Need ultrasonic)
SMILES O=C(O[C@@H](COC1=CN2C(C(OC3=C(C4=C(NC(C)=C4)C=C3)F)=NC=N2)=C1C)C)[C@@H](N)C
Molecular Weight (g/mol) 441.46
Formula Weight 441.46
Percent Purity 98.91%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.