missing translation for 'onlineSavingsMsg'
Learn More

Brilanestrant, MedChemExpress

Product Code. 16475408
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475408 10 mg 10mg
16441597 50 mg 50mg
16428127 5 mg 5mg
16416016 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16475408 Supplier Medchem Express Supplier No. HY12864

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Brilanestrant (ARN-810; GDC-0810) is an orally bioavailable selective estrogen receptor degrader (SERD) with IC50 of 0.7 nM.

Brilanestrant (ARN-810

Specifications

Chemical Name or Material Brilanestrant
CAS 1365888-06-7
Color White
Purity Grade Notes Research
Molecular Formula C26H20ClFN2O2
Quantity 10 mg
Synonym ARN-810 GDC-0810
Solubility Information DMSO : 100 mg/mL (223.76 mM; Need ultrasonic)
SMILES O=C(O)/C=C/C1=CC=C(/C(C2=CC3=C(NN=C3)C=C2)=C(C4=CC=C(F)C=C4Cl)/CC)C=C1
Molecular Weight (g/mol) 446.9
Formula Weight 446.9
Percent Purity 98.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.