Learn More
Branson Ultrasonics™ Bransonic™ Ultrasonic Cleaning Bath Accessory, Perforated Trays
Product Code. 10244584
Click to view available options
Capacity (Metric):
2.27 L
3.4 L
6.8 L
11.4 L
25 L
For Use With (Equipment):
Ultrasonic Bath, Model 1800
Ultrasonic Bath, Model 2800
Ultrasonic Bath, Model 3800
Ultrasonic Bath, Model 5800
Ultrasonic Bath, Model 8800
Unit Size:
Each
Product Code. 10244584
Brand: Branson Ultrasonics™ 100410166
Only null left
Add to basket
Add to basket
For use with Branson ultrasonic baths
  • Accessories for ultrasonic bench-top system
Specifications
Type Perforated Tray
Capacity (Metric) 11.4 L
Capacity (English) 2.5 gal.
For Use With (Equipment) Ultrasonic Bath, Model 5800
Material Stainless Steel
Dimensions (L x W x H) 29.2 x 24.1 x 15.2 cm