Learn More
Branebrutinib, MedChemExpress
Product Code. 16481016
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Product Code. 16481016
Brand: Medchem Express HY112161
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Branebrutinib (BMS-986195) is a highly potent, selective covalent, irreversible inhibitor of Bruton’s tyrosine kinase (BTK), with an IC50 of 0.1 nM.
Branebrutinib (BMS-986195) is a highly potent, selective covalent, irreversible inhibitor of Bruton’s tyrosine kinase (BTK), with an IC50 of 0.1 nM.
Specifications
Chemical Name or Material Branebrutinib
CAS 1912445-55-6
Color White
Purity Grade Notes Research
Molecular Formula C20H23FN4O2
Quantity 200 mg
Synonym BMS-986195
Solubility Information DMSO : ≥ 100 mg/mL (269.96 mM)
SMILES O=C(C1=CC(F)=C(N2C[C@@H](NC(C#CC)=O)CCC2)C3=C1NC(C)=C3C)N
Molecular Weight (g/mol) 370.42
Formula Weight 370.42
Percent Purity 99.52%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease