Learn More
VACUUBRAND™ MD 4 NT Diaphragm Vacuum Pump
GREENER_CHOICE
Product Code. 10090113
Click to view available options
Flow Rate:
2.2/2.5 CFM
Unit Size:
Each
Product Code. 10090113
Brand: VACUUBRAND™ 20736000
Only null left
Add to basket
Add to basket
For a wide variety of non-corrosive and non-evaporative applications. MD-series pumps an ultimate vacuum of 1.1 Torr.
Utilizes diaphragm vacuum technology for totally dry operation; there is no oil to change or monitor.
  • Minimizes downtime and service costs
  • Unique design eliminates tedious, trial-and-error stroke length recalibration
  • Individually accessible diaphragms and valves simplify servicing
  • Feature specially engineered pump heads for high flow rates at working vacuum
TRUSTED_SUSTAINABILITY
Specifications
Max. Vacuum 0.75 torr
Horsepower 0.34 hp
Amperage 3 A
Includes Manual
Motor Speed 150 to 1800 rpm
Certifications/Compliance ATEX Conformity II 3G IIC T3 X, CE, DIN EN 1012-2:2011, DIN EN 12100:2001, DIN EN 50581:2013, DIN EN 61010-1:2011, DIN EN 61326-1:2013, IEC 61010-1:2010, NRTL Certification, TUV
Electrical Requirements 230 V 50/60 Hz
Dimensions (L x W x H) 12.7 x 9.4 x 7.7 in. (32.5 x 23.8 x 19.8 cm)
Material Aluminum Alloy (Case)
Model MD 4 NT
Noise Level 45 dB(A)
Plug Type CEE
Temperature (Metric) Storage -10°C to +60°C
Weight (English) 36.1 lb.
Weight (Metric) 16.4 kg
Temperature (English) Max. 104°F
Temperature (Metric) Max 40°C
Temperature (Metric) Min. 10°C
Flow Rate 2.2/2.5 CFM
Frequency 50/60 Hz
Overall Height (English) 7.7 in.
Overall Height (Metric) 198.12 mm
Overall Length (English) 12.7 in.
Overall Length (Metric) 325.12 mm
Overall Width (English) 9.4 in.
Overall Width (Metric) 23.8 cm
Pump Type Diaphragm Pump
Enclosure Rating IP40
No. of Pump Heads 4
Temperature (English) Min 50°F
Applications Non-Corrosive Gases
Oil Type Oil-Free
No. of Stages 3
Maximum Pressure 16 psi
Voltage 230 V