Learn More
VACUUBRAND™ Supplementary Kit Connection for BVC Fluid Aspiration Systems
Click to view available options
For Use With:
BVC fluid aspiration system
Unit Size:
1 set
Description
The BVC family of fluid aspiration systems deliver compact and quiet solutions for cell culture work. They provide the flexibility to accommodate decontamination protocol whether autoclaving or disinfecting in the bottle.
- To add second VHCPRO to BVC bottle aspiration cap
Specifications
Specifications
| For Use With | BVC fluid aspiration system |
| Type | Supplementary Kit Connection |