Learn More
BRAND™ Self-adhesive Plate Sealing Film
Product Code. 10767564 Shop All Brand Products
Click to view available options
Material:
Aluminum
Polyester
Polyethylene, Polypropylene
Polypropylene
Rayon
Vinyl
For Use With (Application):
Automation
Cell and tissue culture
Cold storage
ELISA, PCR
ELISA, Real-time PCR
Luminescence measurements
Storage
Storage, fluorescence measurements
No. per Pack:
100
100 Sheets
50 Sheets
Unit Size:
Pack of 100
Pack of 50
Product Code. 10767564
Brand: BRAND™ 781391
Securely seals multiwell plates. Self-adhesive Plate Sealing Film can be applied directly to plates and removed without the use of expensive equipment, or can be applied with a hard rubber roller to ensure secure and uniform attachment of the film to the plate.
  • Easily apply to plates and remove without the use of expensive equipment.
  • Application with a hard rubber roller will ensure secure and uniform attachment of the film to the plate.
  • Sealing films are available in different materials especially suited for storage, immunoassay, or cell and tissue culture:
    • Polyethylene
    • Polyester
    • Aluminum
    • Rayon
    • Polypropylene
    • Vinyl
TRUSTED_SUSTAINABILITY
Specifications
Description Highly transparent
Material Polyester
No. per Pack 100 Sheets
For Use With (Application) ELISA, Real-time PCR
Product Type Sealing Film
Temperature Range (Metric) -40°C to +110°C