Learn More
BRAND™ 384-Well PCR Plates
Shop All Brand Products
Click to view available options
Product Type:
Full skirt
Unit Size:
Pack of 50
Description
- Non-sterile, 0.3mL BRAND™ 384-well deep well plate has a square bottom and is manufactured from high-quality virgin polypropylene
- Excellent for high throughput screening (HTS), assays requiring a mother plate, cell and tissue culture, sample storage, and more
- Plate is resistant to a wide variety of chemicals, including phenols, chloroform, and DMSO and is compatible with most robotic samplers and automated liquid handling systems
- Plate withstands temperatures as low as -121ºC, making it an excellent choice for cold-room applications and cryo-storage
- PP plate is autoclavable at 250ºF (121ºC) for 20 minutes
- Wells are labeled in a standard alpha-numeric pattern to simplify sample identification, and have notched corners to facilitate orientation
Specifications
Specifications
| Format | Standard |
| Array | 16 x 24 |
| Autoclavable | Autoclavable |
| Product Type | Full skirt |
| Skirt Style | Full |
| Material | Polypropylene |
| No. of Wells | 384 |
| For Use With (Application) | qPCR |
| Quantity | 50/Pk. |