missing translation for 'onlineSavingsMsg'
Learn More

Scharlab Brain Heart Infusion Broth

Product Code. 11411767
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411767 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11411767 Supplier Scharlab Supplier No. 02599500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Brain Heart Infusion Broth (BHI Broth) is used for the cultivation of fastidious bacteria and pathogenic fungi.

  • BAM/COMPF/ISO/SMWW
  • Nutrient rich medium suitable for the isolation of pathogenic microorganisms from clinical specimens
  • According to ISO 5944 standard

Specifications

Certifications/Compliance ISO 5944
Form Liquid
Media Type Brain Heart Infusion Broth
Product Type Microbiology Media
Target Organism Class Fastidious
Formulation Enzymatic Digest of Animal Tissues, and Brain-Heart Extract 27.5 g/L, Dextrose 2.0 g/L, Sodium Chloride 5.0 g/L, Disodium Phosphate (Anhy.) 2.5 g/L
For Use With (Application) Isolation of Pathogenic Microorganisms
Incubation Conditions 37°C ± 1.0°C
Incubation Time 24 ±2 hr.
pH 7.4 ±0.2
Quantity 500 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.