Learn More
Brady™ Printer i5100 - UK/EU with Brady Workstation LAB Suite
Click to view available options
Resolution:
300 dpi
600 dpi
Description:
With Cutter and Software
With Software
Unit Size:
1 set
Description
- The BradyPrinter i5100 Industrial Label Printer offers a perfect mix of performance & smarts
- This benchtop printer can print on 70 label materials, more than 1200 stock label parts and tailored custom label materials and parts
- Applications include a wide range of specialised and commonly used product labels, electronics and PCB labels, logistics and asset tracking labels, cable & component identification labels and laboratory sample labels
- The printer comes with the Brady Workstation Laboratory Identification Software suite enabling you to immediately start designing professional labels
- This suite combines frequently used software apps to create laboratory identification labels.
Specifications
Specifications
| Display Type | Color Touchscreen |
| Print Technology | Thermal Transfer |
| Width (Metric) | 240.00 mm |
| Height (Metric) | 317.00 mm |
| Depth (Metric) | 435.00 mm |
| Dimensions (L x W x H) | 435.00 x 240.00 x 317.00 mm |
| Color Capability | Monochrome |
| Description | With Cutter and Software |
| Includes | Power cord, a USB cable, a product CD holding the driver, the user manual in languages, the configuration/settings manual in English and a printed user manual in English, an RFID enabled roll holder, a ribbon take up spindle and a Brady Workstation Laboratory Identification Suite |
| Print Speed | 150.00 mm/s |
| Show More |