Learn More
Brady™ BradyPrinter i5100 Kit UK and EU with Workstation PWID Suite
Product Code. 16429253
Click to view available options
:
BradyPrinter i5100; Color Capability: Monochrome; Print Speed: 150 mm/s; Print Technology: Thermal Transfer; Resolution: 600 dpi; Weight (Metric): 6.2 kg
Product Code. 16429253
Brand: Brady™ 198780
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The BradyPrinter i5100 Industrial Label Printer is smart and easy to use, with excellent performance and the ability to print more than 1200 standard identification labels, plus custom labels.
Create and print professional labels with the BradyPrinter i5100 Kit. Intelligent Print (IP) technology allows the materials, printer and included Brady™ Workstation PWID Suite software to communicate.
  • Change printer settings easily with intuitive touchscreen
  • Change out materials quickly with specially designed materials that easily lock into place
  • Connect easily with a variety of connection ports
  • Keep printer running smoothly with interchangable rollers
  • Prints more than 1200 standard identificaiton labels plus custom labels
  • Includes Brady™ Workstation Product and Wire Identification Software Suite
Specifications
Display Type Color LCD Touchscreen
Print Technology Thermal Transfer
Width (English) 9.5 in.
Width (Metric) 241 mm
Height (English) 12.5 in.
Height (Metric) 318 mm
Depth (English) 17 in.
Depth (Metric) 437 mm
Autoclavable Not Autoclavable
Sterility Non-sterile
Color Capability Monochrome
Includes i5100 Printer, Power Cord, USB Cable, Product CD with Driver, User Manual, RFID Enabled Roll Holder, Ribbon Take Up Spindle, Brady™ Workstation PWID Suite
Languages Arabic, Bulgarian, Chinese (Tradditional), Chinese (Simplified), Croatian, Czech, Danish, Dutch, English, Finnish, French, German, Greek, Hungarian, Italian, Lithuanian, Norwegian, Polish, Portuguese, Romanian, Russian, Serbian, Slovenian, Spanish, Swedish, Turkish
Memory 256 MB RAM, 50 MB IFFS Flash, SD Card, Slot Up To 512 GB
Model i5100
Print Speed 150 mm/s
Resolution 600 dpi
Connector Types Bluetooth™, Ethernet, Serial, USB
Operating System Windows™ 10, Windows™ 7, Windows™ 8, Windows™ Servers 2012, 2012 R2, and 2016
Temperature Range (Metric) 0°C to 40°C
Weight (English) 14 lb.
Weight (Metric) 6.2 kg
For Use With (Application) Asset Tracking, Circuit Board Labeling, General Identification
Warranty 2 Year
Voltage 110/240 V